Imprimer
Stopcocks, straight bore
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SSD102 SSD103 SSD104
BIBBSSD102EA 19.9 EUR
BIBBSSD102 BIBBSSD103 BIBBSSD104
Stopcocks, straight bore
Robinets d'arrêt
Straight bore stopcock, made of borosilicat glass 3.3, with plain arm and interchangeable glass key.
  • Glass stopcocks which meet the specifications of DIN 12541
  • Leakage rate meets the requirements of DIN 12540
Informations de livraison: Supplied complete with key retaining device.