Imprimer
Dropping funnels, cylindrical, without pressure balance
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
443333 443340 443347 443354 443360
BIBB443333EA 125 EUR
BIBB443333 BIBB443340 BIBB443347 BIBB443354 BIBB443360
Dropping funnels, cylindrical, without pressure balance
Entonnoirs Ampoules de coulée
Dropping funnel, cylindrical shape, made of Pyrex® borosilicate glass, with cone and socket. Stopcock with interchangeable PTFE key.
  • Excellent chemical and temperature resistance
  • White enamel double graduated scale
  • Stem inside cone for easy fitting to Quickfit® assemblies
Certifications: Complies with ISO 4800.