Imprimer
Screw caps for laboratory bottles
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4506/25D 4506/32D 4506/45D 4506/45E 4506/45F 4506/45G 4506/80D
BIBB4506/25DEA 35.1 EUR
BIBB4506/25D BIBB4506/32D 215-3136 BIBB4506/45E BIBB4506/45F BIBB4506/45G BIBB4506/80D
Screw caps for laboratory bottles
Capuchons Bouchons pour flacons
Screw cap, made of PP, for media-lab bottles
  • Plug seal cap ideal for media preparation work
  • Autoclavable up to 140°C
  • Available in choice of 4 colours for easy identification of samples