Imprimer
Flacons PYREX®, Media-lab, transparents, sans bouchon ni bague anti-goutte
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1517/01 1517/02 1517/04 1517/06 1517/08 1517/10 1517/12 1517/16 1517/18 1517/05 1517/14
BIBB1517/01EA 83.2 EUR
BIBB1517/01 215-3528 215-3129 215-3130 215-3529 215-3530 215-3535 215-3536 BIBB1517/18 BIBB1517/05 BIBB1517/14
Flacons PYREX®, Media-lab, transparents, sans bouchon ni bague anti-goutte
Flacons Flacons en verre
These media-lab bottles are the same as the 1516 range, but are supplied without the blue polypropylene screw cap and clear pouring ring.
  • Bottles only; supplied without cap or pouring ring
  • Printed with Trace Code for downloadable batch certificate
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
Manufactured from temperature-resistant and chemically-resistant borosilicate glass, these bottles are ideal for mixing and storage of culture medium.
The 25 ml size bottle features a glass lip and does not require a pouring ring.
Certifications:
Complies with ISO 4796.