Imprimer
Bottles, narrow neck, round, HDPE, with screw caps, Azlon®
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BNH2000P BNH0030PN BNH0060PN BNH0125PN BNH0250P BNH0500P BNH1000P BNH0100PN
215-4055EA 68 EUR
215-4055 215-3359 215-4054 215-3100 215-3105 215-3107 215-3847 215-3360
Bottles, narrow neck, round, HDPE, with screw caps, Azlon®
Flacons Flacons en plastique
HDPE, translucent with PP screw cap.
  • Excellent general chemical resistance
  • Heat resistance from –100 to +120 °C
  • Lightweight but very durable in use
Ideal for storing a wide range of liquids.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904