Imprimer
Stirring rods, glass
Fournisseur: SciLabware
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SRF380 SRF384 SRF386
BIBBSRF380EA 52.9 EUR
BIBBSRF380 441-0137 441-0138
Stirring rods, glass
Agitateurs Tiges d'agitation
Stirring rods with flattened end forming a paddle 20×9 mm at one end, and flattened smoothed top approximately 9 mm in diameter, which can be used to crush lumps.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable