Imprimer
Colonnes capillaires, OPTIMA® 225
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
726080.10 726118.15 726118.25 726118.30 726118.50 726118.60 726352.25 726352.30 726352.50 726352.60
MANA726080.10EA 478 EUR
MANA726080.10 MANA726118.15 MANA726118.25 MANA726118.30 MANA726118.50 MANA726118.60 MANA726352.25 MANA726352.30 MANA726352.50 MANA726352.60
Colonnes capillaires, OPTIMA® 225
Colonnes pour chromatographie
Medium polar GC phase.
  • Recommended for fatty acid analyses
  • Maximum temperature for isothermal operation 260 °C
  • Maximum temperature for short isotherms in a temperature program 280 °C
50% cyanopropylmethyl – 50% phenylmethylpolysiloxane phase.
USP code: G7 / G19.