Imprimer
Magnetic stirring bars, PTFE-coated, octagonal, with centre ring
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
42077008
HECH42077008EA 12 EUR
HECH42077008
Magnetic stirring bars, PTFE-coated, octagonal, with centre ring
Agitateurs Barreaux magnétiques
PTFE coated, magnetic stirring bars.
  • Pivot ring reduces surface contact and ensures stirrer is in optimum position in the vessel.