Imprimer
PCR tubes, with flat or domed cap, Sapphire
Fournisseur: Greiner Bio-One
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
683201_1000 671201 671273_1000 671275_1000 671276_1000 671277_1000 682201_1000 683271_1000
GREI683201_1000EA 114 EUR
GREI683201_1000 GREI671201 GREI671273_1000 GREI671275_1000 GREI671276_1000 GREI671277_1000 GREI682201_1000 GREI683271_1000
PCR tubes, with flat or domed cap, Sapphire
Tubes Tubes PCR
Manufactured out of medical-grade polypropylene without interfering additives.
  • Thin-walled for efficient heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Suitable for real-time / quantitative PCR
  • Comprehensive thermal cycler compatibility
Polypropylene tubes feature thin-wall construction for optimal heat transfer from the thermocycler block to the reaction mixture.