Imprimer
Cap sealing mats, Matrix SepraSeal® and DuraSeal®
Fournisseur: THERMO MATRIX TECHNOLOGIES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4465 4464 4466 4490 4463BLU 4463GRE 4463YEL 4463PUR 4463GRY 4463RED 4464GRE 4464YEL 4464BLU 4464RED 4464GRY 4464PUR 4465BLU 4465GRE 4465RED 4465PUR 4465YEL 4466BLU 4466GRE 4466GRY 4466PUR 4466RED 4466YEL 4463
479-1246EA 147 EUR
479-1246 479-1245 479-1247 MATR4490 479-1267 479-1268 479-1272 479-1270 479-1269 479-1271 479-1274 479-1278 479-1273 479-1277 479-1275 479-1276 479-1279 479-1280 479-1284 479-1283 479-1285 479-1286 479-1287 479-1288 479-1289 479-1290 479-1291 MATR4463
Cap sealing mats, Matrix SepraSeal® and DuraSeal®
Scellés Microplaque Sceaux
SepraSeal® and DuraSeal® capping systems offer a pierceable, individual tube sealing solution. They are made from thermoplastic elastomer (TPE) with colorants and are DMSO resistant.
  • Low vapour transmission rates minimises evaporation and water uptake
  • Pierceable design allows samples to be accessed without removing caps
  • TPE construction ensures sealing performance will not degrade, even after multiple piercings
The SepraSeal® capping system offers the flexibility of individual tube capping plus the ability to apply 96 caps at once. Designed for storage down to −80 °C, SepraSeals® are available in a variety of colours in both sterile and non sterile configurations, with solid or pre-split cap designs. To access samples, solid seals can be removed with the SepraSeal® cap removal tool or pierced by liquid handling needles. For repetitive sample access, a pre-split SepraSeal® can be pierced multiple times with very little force required. Seal integrity is maintained due to its self-closing nature.
DuraSeal® is a permanently-installed, ultra-robust version of a SepraSeal® that enables storage down to the vapour phase of liquid nitrogen. It is ideal for applications requiring repetitive piercings. The V-shape of the DuraSeal® reliably guides liquid handling needles to the same location repeatedly with same self-healing capability as a SepraSeal®.