Imprimer
Flasks acc. to Kjeldahl
Fournisseur: Kavalier
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
632411210050 632411210250 632411210500 632411210100
214-1341EA 20.1 EUR
214-1341 214-1342 214-1343 214-1345
Flasks acc. to Kjeldahl
Ballons Flacons Kjeldahl
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With printed indication of contents.