Imprimer
Soil analysis filter papers, phosphate-free
Fournisseur: MACHEREY-NAGEL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
483009 483011 521015 540015 540018 521011 540012 440005 440007 440009 440011 440012 440015 440018 440024 440027 440032 440038 440040 440045 440050 483005 483007 483012 483015 483018 483024 483027 483032 483038 483040 483045 483050 494005 494007 494009 494011 494012 494015 494018 494024 494027 494032 494038 494040 494045 494050 521012 521018 521024 540005 540007 540009 540011 540024 540027 540032 540038 540040 540045 540050
MANA483009EA 16 EUR
MANA483009 MANA483011 MANA521015 MANA540015 805-4018 516-5008 MANA540012 MANA440005 MANA440007 MANA440009 MANA440011 MANA440012 MANA440015 MANA440018 MANA440024 MANA440027 MANA440032 MANA440038 MANA440040 MANA440045 MANA440050 MANA483005 MANA483007 MANA483012 MANA483015 MANA483018 MANA483024 MANA483027 MANA483032 MANA483038 MANA483040 MANA483045 MANA483050 MANA494005 MANA494007 MANA494009 MANA494011 MANA494012 MANA494015 MANA494018 MANA494024 MANA494027 MANA494032 MANA494038 MANA494040 MANA494045 MANA494050 MANA521012 MANA521018 MANA521024 MANA540005 MANA540007 MANA540009 MANA540011 MANA540024 MANA540027 MANA540032 MANA540038 MANA540040 MANA540045 MANA540050
Soil analysis filter papers, phosphate-free
Filtres Papiers filtre Papiers filtres qualitatifs
Phosphate-free smooth filter papers for soil analysis.
MN 280 1/4 folded filters made from acid-washed paper with a high clarification efficiency for determination of micro-nutrients available to vegetation.
MN 619 G for slow filtration, MN 616 G for medium fast filtration and MN 617 G for fast filtration.