Imprimer
Special tweezers, coated
Fournisseur: RSG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
416.110 416.130
RSGA416.110EA 144 EUR
RSGA416.110 RSGA416.130
Special tweezers, coated
Pinces
These special tweezers made of stainless steel feature a dark PTFE coating.
  • Extraordinarily fine
  • Precision cut tips