Cinchophène ≥99%
Marques : Thermo Fisher ScientificTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
|
Danger
|
Synonyme(s):
2-Phenyl-4-quinolinecarboxylic acid, Cinchophen
Purity: 99%
CAS No.: 132-60-5
Molecular Formula: C16H11NO2
Formula Weight: 249.27
Odour: Odourless
Melting Point: 212-218°
Beilstein Reference: 192803
Merck Reference: 14,2288
MDL No.: MFCD00006750
Formule:
C₁₆H₁₁NO₂ Point de fusion: 212…218 °C Température de stockage: Température ambiante |
Numéro MDL:
MFCD00006750 Numéro CAS: 132-60-5 EINECS: 205-067-1 Indice Merck: 14,2288 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...