Imprimer
Distributeurs adaptables sur flacon, Finnpipette®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4421120 4421130 4421140 4421150 4421160 4421170
LABS4421120EA 457 EUR
LABS4421120 LABS4421130 LABS4421140 LABS4421150 LABS4421160 LABS4421170
Distributeurs adaptables sur flacon, Finnpipette®
Distributeurs pour flacons
This bottle top dispenser feature a superior tool for dosing liquids from reagent bottles that have six models available.
  • Ergonomically designed, non-slip piston handle
  • Excellent chemical and thermal resistance
  • Volumes are absolutely reproducible
  • Threaded suction hose for bubble free suction
They can be fully autoclaved without disassembling and have precise self-locking system in the volume setting.