Imprimer
Concentrator
Marques : G Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-144 786-143
786-144EA 225 EUR
786-144 786-143
Concentrator
Protéines purification Protein Purification Dialysis
The concentrator is a high molecular weight polymer which will not migrate across the dialysis membrane. The concentrator is placed against the dialysis membrane of the Tube-O-DIALYZER™, where it rapidly absorbs water from the sample and reduces the sample volume. The concentrator can be used with dialysis bags and other dialysis devices.