Imprimer
Device for protein cross linking, coupling and modification, Tube-O-Reactor™
Fournisseur: G-Biosciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
786-027-15K 786-027-4K 786-027-8K 786-024-15K 786-024-4K 786-024-8K
786-027-15KEA 258 EUR
786-027-15K 786-027-4K 786-027-8K 786-024-15K 786-024-4K 786-024-8K
Device for protein cross linking, coupling and modification, Tube-O-Reactor™
Tubes Tubes de dialyse
Tube‐O‐Reactor™, Micro, for 20 to 250 µl samples or Medi for 0,2 to 2 ml samples.
The Tube‐O‐Reactor™ is a simple reactor-device for performing a wide variety of reactions including crosslinking, modification, and labeling of proteins and nucleic acids. Most reactions involve three critical steps ‐ equilibration of reaction conditions for efficient reaction, reaction with target specific agents, and then removal of un-reacted agents and bi‐products, followed by recovery of the reaction products. Tube‐O-Reactor™ has been designed to perform all three critical steps ofEn savoir plus