Imprimer
Test tubes, flat bottom, without rim, heavy wall
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
42779070 42779065 92277901450
HECH42779070EA 61.4 EUR
HECH42779070 HECH42779065 HECH92277901450
Test tubes, flat bottom, without rim, heavy wall
Tubes Tubes à essais
Made of AR-Glas® (soda glass).
  • Heavy wall
  • Flat bottom
  • Good chemical and physical properties