Imprimer
Separating funnels, conical
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
42951030 42951024 42951060 42950024 42950030 42950048 42950060 42950066 42950072
HECH42951030EA 124 EUR
HECH42951030 HECH42951024 HECH42951060 HECH42950024 HECH42950030 HECH42950048 HECH42950060 HECH42950066 HECH42950072
Separating funnels, conical
Entonnoirs Ampoules à décanter
Separating funnels, conical, made of DURAN® borosilicate glass 3.3, with solid glass or interchangeable PTFE key and plastic stopper.
  • Large labeling field
  • Excllent temperature resistance
  • Excllent chemical resistance
Certifications: DIN ISO4800