Imprimer
LC-MS nitrogen generators
Fournisseur: PerkinElmer
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N9306211 N9306214
PERCN9306211EA 59200 EUR
PERCN9306211 PERCN9306214
LC-MS nitrogen generators
Générateurs de gaz Générateurs d'azote
Nitrogen generators are designed to supply single or multiple LC-MS instruments with dry nitrogen at purities of 98 – 99,5%. The generator can also be used for solvent evaporation as well as supplying dry nitrogen to analytical instruments.