Imprimer
Chemical resistant overalls, Mutex Light+
Fournisseur: HONEYWELL SÉCURITÉ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4500501/XL 4500501/XXL 4500501/XXXL 4500501/M 4500501/S
DALL4500501/XLEA 0 EUR
DALL4500501/XL DALL4500501/XXL DALL4500501/XXXL DALL4500501/M DALL4500501/S
Chemical resistant overalls, Mutex Light+
Vêtements à usage unique
These disposable overalls made of PP laminated with a microporous PE film are designed to protect against particles contamination and chemical splashes.
  • Elasticated 3-piece hood, waist, wrists and ankles
  • Special crotch design
  • Outside flap and dual slider zipper
  • Silicone and latex-free
  • Anti-static and radioactive particles certified
Typical applications include: Mechanical works, maintenance on site, industrial cleaning in wet environments.
Weight: 65 g/m².
Certifications: Type 5 + 6; Cat. III