Imprimer
Syringes for HPLC autosamplers, for WATERS® WISP™
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
80024 80020
549-1253EA 273 EUR
549-1253 549-1252
Syringes for HPLC autosamplers, for WATERS® WISP™
Seringues Seringues de chromatographie
These gas tight syringe are designed for HPLC autosamplers.
  • Temperature range: 10 to 115 °C