Imprimer
Gas inlet valves, VGS and VGL
Fournisseur: VACUUBRAND
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20666000 20666400
183-1211EA 310 EUR
183-1211 183-1212
Gas inlet valves, VGS and VGL
Vannes Vannes à vide
Gas inlet valves are small hand operated valves for admitting gases into a vacuum system.
The vacuum and atmospheric sides of the valve VGL are provided with hard-soldering brass connections, facilitating the valve to be used as a fixed, permanent fitting.