Imprimer
High vacuum bellow valves, VE series
Fournisseur: VACUUBRAND
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20664004 20664005 20664006 20674020 20674021 20674022
VACU20664004EA 462 EUR
VACU20664004 VACU20664005 VACU20664006 VACU20674020 VACU20674021 VACU20674022
High vacuum bellow valves, VE series
Vannes Vannes à vide
High vacuum, helium leak-tested right angle valves VE have stainless steel bellows which seals the drive mechanism without any gap on the vacuum side.
  • High conductance due to large stroke length
  • Excellent gas tightness, minimum degassing
  • Stainless steel version VE WIG bakeable to 150 °C
  • Simple dismantling and easy changing of seal
  • Dimensions of the VE valve are the same as elbow with corresponding nominal width