Imprimer
Flasks, round bottom, with standard ground joints
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
41913010 41913014 41913012 41913018 41913024 41913030
HECH41913010EA 33.4 EUR
HECH41913010 HECH41913014 HECH41913012 HECH41913018 HECH41913024 HECH41913030
Flasks, round bottom, with standard ground joints
Ballons Flacons à fond rond
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.