Imprimer
Rubbermaid® Heavy Duty Carts with Lipped Shelves
Fournisseur: Rubbermaid Commercial Products
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FG452088BLA FG450088BLA
RUBBFG452088BLAEA 671 EUR
RUBBFG452088BLA 129-1829
Rubbermaid® Heavy Duty Carts with Lipped Shelves
Chariots
The BRUTE Heavy Duty Utility Cart with lipped shelves is a durable versatile cart that is ideal for transporting materials, supplies, and heavy loads up to 226 kg.
  • Premium ergonomic handle provides optimal hand placement to improve control
  • Features two 12.7 cm non-marking 2 swivel casters and 2 locking casters that keeps the cart in place when loading and unloading
  • Durable structural foam construction is lightweight and maneuverable with 226 kg of storage weight capacity
  • Lipped shelf features integrated V-notch to hold pipe and conduit securely for safe cutting
  • Built-in tool holster and caddy keep items at hand and in place during transport
  • Molded-in fittings allow you to add pipe or rods to hold wire spools and other roll-type goods
  • Easy-reach tool/accessory hooks provide additional storage
Features two 127 mm non-marking 2 swivel casters and 2 locking casters that keeps the cart in place when loading and unloading.
Certifications: Locking casters compliant with EN 1757-3.