Imprimer
Containers with screw cap, Sisterile®
Fournisseur: KIMA SAS DI CHIARIN RENZO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
231180 231196
KIMA231180EA 308 EUR
KIMA231180 KIMA231196
Containers with screw cap, Sisterile®
Récipients de collecte d'échantillon Conteneurs à usage universel
Made of PP, translucent.
  • Graduated
  • Excellent chemical resistance
  • Very good temperature resistance
Labelled with writing surface of 28×48 mm and screw cap.