Imprimer
Jars with base, knobbed lid
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
44410024 44410066 44410072 44410030 44410036 44410042 44410018 44410054 44415018 44415054 44415024 44415066 44415072 44415030 44415036 44416036
216-4293EA 80.6 EUR
216-4293 216-4294 HECH44410072 216-4295 HECH44410036 216-4296 HECH44410018 216-4297 HECH44415018 HECH44415054 HECH44415024 216-4298 HECH44415072 216-4299 HECH44415036 HECH44416036
Jars with base, knobbed lid
Jars Specimen Jars
Made of glass, with glass or stainless steel lid.
  • Good chemical resistance
  • Sturdy
  • Easy to clean
Suitable for storing dressing material, cotton, clinical thermometers, forceps and other instruments.