Imprimer
Urine container, with collection device for vacuum test tubes
Fournisseur: KIMA SAS DI CHIARIN RENZO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
21800 21820 21780 21883 21870 21890 21870E
720-2862EA 300 EUR
720-2862 KIMA21820 720-2861 KIMA21883 KIMA21870 KIMA21890 KIMA21870E
Urine container, with collection device for vacuum test tubes
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons d'urine
Sterile containers for urine culture or non sterile for physical chemical microscopic analysis.
  • Collection spot with a partially removable adhesive label to be reclosed after the collection
  • The labels are printed with patient and operators instructions