Imprimer
Filter centering rings for small flange connections
Fournisseur: VACUUBRAND
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20660160 20660161 20660162 20660163
185-1291EA 46 EUR
185-1291 185-1292 185-1293 185-1294
Filter centering rings for small flange connections
Scellés Bagues d'étanchéité
These filter centering rings for small flange KF connections made of stainless steel with NBR sealing ring.