Imprimer
Ground joint adaptors, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
H980-06 H980-03 H980-09 H980-12
BOHLH980-06EA 56.3 EUR
BOHLH980-06 BOHLH980-03 BOHLH980-09 BOHLH980-12
Ground joint adaptors, BOLA
Adaptateurs Raccords à rodage conique
PTFE, white. For connecting different ground joint sizes.
  • Temperature resistance: –200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
Socket in cone, with sealing rings on the outside and knurled grip.