Imprimer
HPLC system, Primaide™
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
903-0700EA 12600 EUR
903-0700 903-0701 903-0702 903-0703 903-0704 903-0705 903-0706 903-0709 903-0707 903-0798 903-0799 903-0741
HPLC system, Primaide™
Chromatography Systems Complete HPLC Systems
The Primaide™ is a standard HPLC system designed and manufactured to the highest quality by Hitachi. Suitable for academic and other small laboratories that need a sturdy and reliable standard HPLC system.
  • Flexible modular system
  • Auto calibrating UV and DAD detectors
  • Easy to use and simple software
  • Direct injection autosampler
  • Highly robust and proven technology
Informations de livraison: All main system modules include an accessory set, an E-line cable, instruction manual, validation certificate and test report.