Imprimer
Miniature screw-in adaptors, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F716-08 F716-06 F716-04 F716-02
BOHLF716-08EA 71.1 EUR
BOHLF716-08 BOHLF716-06 BOHLF716-04 BOHLF716-02
Miniature screw-in adaptors, BOLA
Raccords et connecteurs
Adaptor made of PTFE for transition from female thread UNF ¼" 28 G to male thread NPT ¹/₈" or NPT ¼".
  • Temperature resistance: from –200 to +250 °C
  • Universal chemical resistance