Imprimer
Filtering flasks with glass hose connection
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
41270036 41270042 41270048 41270054
HECH41270036EA 142 EUR
HECH41270036 HECH41270042 HECH41270048 HECH41270054
Filtering flasks with glass hose connection
Ballons Fioles à vide
Flasks for vacuum use, made of DURAN® borosilicate glass.
  • Erlenmeyer shape
  • Olive hose connection Ø 11 mm
  • Does not conform to the 'Equipment and product safety regulations'
ISO 6556