Imprimer
Vacuum desiccators, Novus
Fournisseur: GLASWARENFABRIK KARL HECHT
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
41160010 41160015 41160020 41160025 41160030
HECH41160010EA 273 EUR
HECH41160010 HECH41160015 HECH41160020 HECH41160025 HECH41160030
Vacuum desiccators, Novus
Dessiccateurs Dessiccateurs en verre
Vacuum desiccators are made of borosilicate glass 3.3 and are tested for tension, vacuum tightness and pressure.
Certifications: DIN 12491.