Imprimer
Measuring cylinders, tall form, class A, borosilicate glass, DAkkS calibrated, BLAUBRAND®
Fournisseur: Brand
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
DAKKS32138 DAKKS32148 DAKKS32164 DAKKS32162 DAKKS32154 DAKKS32120 DAKKS32128 DAKKS32108 DAKKS32105
BRNDDAKKS32138EA 109 EUR
BRNDDAKKS32138 BRNDDAKKS32148 BRNDDAKKS32164 BRNDDAKKS32162 BRNDDAKKS32154 BRNDDAKKS32120 BRNDDAKKS32128 BRNDDAKKS32108 BRNDDAKKS32105
Measuring cylinders, tall form, class A, borosilicate glass, DAkkS calibrated, BLAUBRAND®
Cylinders Measuring Cylinders
Borosilicate 3.3 glass, tall form, class A, conformity certified.
  • Includes DAkkS calibration certificate
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, with spout
  • Ring marks at major graduations, marks and inscriptions in high contrast blue enamel
Certifications: DIN EN ISO 4788