Imprimer
Beakers, low form, with spout
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
8.600.100 8.600.050 8.600.150
WITG8.600.100EA 0 EUR
WITG8.600.100 WITG8.600.050 WITG8.600.150
Beakers, low form, with spout
Béchers
Quartz glass.
  • High thermal shock resistance with low thermal expansion
  • High transmission from the UV to the IR spectrum range
  • High chemical resistance and purity
  • High electrical insulation properties
  • High radiation resistance
  • High temperature resistance and softening temperature
The combination of the special thermal, electrical, chemical, mechanical and optical properties in a single material makes quartz glass indispensible for high-technology applications.