Imprimer
Culture flasks, Erlenmeyer shape, BIOGEN
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5.560.250 5.560.050
WITG5.560.250EA 308 EUR
WITG5.560.250 WITG5.560.050
Culture flasks, Erlenmeyer shape, BIOGEN
Ballons Flacons de culture
Culture flask, made of borosilicate glass, Erlenmeyer shape, with straight neck for metal caps.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes