Imprimer
Tissue grinders, Potter-Elvehjem type
Fournisseur: witeg Labortechnik
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5.553.734 5.553.735 5.553.738 5.553.737
WITG5.553.734EA 127 EUR
WITG5.553.734 WITG5.553.735 WITG5.553.738 WITG5.553.737
Tissue grinders, Potter-Elvehjem type
Homogénéisateurs Broyeurs pour tissus
Tissue grinders are made of borosilicate glass, with PTFE pestle and stainless steel shaft.
Clearance between pestle and tube is 102 to 152 μm.