Imprimer
Miniature inert valves, HV series
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
86725 86726 86727 86728 86729 86730 86731 86732 86735 86736 86738 86788 86779 86777 86901 86903 86915 86778 86781 86775 86780 86918 86785 86786 86787 86902 86905 86919 86907 86910 86911 86912 86913 86916 86904
HAMI86725EA 167 EUR
HAMI86725 HAMI86726 HAMI86727 HAMI86728 HAMI86729 HAMI86730 HAMI86731 HAMI86732 HAMI86735 HAMI86736 HAMI86738 HAMI86788 HAMI86779 HAMI86777 HAMI86901 HAMI86903 HAMI86915 613-1617 HAMI86781 HAMI86775 HAMI86780 HAMI86918 HAMI86785 HAMI86786 HAMI86787 HAMI86902 HAMI86905 HAMI86919 HAMI86907 HAMI86910 HAMI86911 HAMI86912 HAMI86913 HAMI86916 HAMI86904
Miniature inert valves, HV series
Vannes Micro-vannes
HV plug valves provide an economical and flexible method of custom-building a fluid flow control system, from a simple on/off valve to a more complex multi-valve arrangement, using a combination of loop and distribution valves.
  • Wetted paths are chemically inert PTFE and Kel-F® materials
  • Valve position is controlled manually via a handle attached to the valve stem
  • Valve can be chosen by selecting the proper flow path
The small valve body with its chemically inert flow path is ideally suited for low pressure applications. All HV Plug Valves allow you to select and interchange a wide variety of fittings, adapters and tubing, including male and female luers, FEP tubing assemblies, and adapters which thread directly into the valve providing an endless variety of valve configurations for any flow application. HV fittings and adapters are installed finger-tight (10 ounce inches) and provide a leak-tight seal tested to 100 psig.
The HV valve is physically the smallest manual valve and constitutes a basic unit for the control of fluids. It is ideal for low pressure applications; from a basic on/off flow to a four-way distribution system.
The HVP valve is essentially the same as the manual HV valve except the HVP can be mounted into panels allowing for custom installation into control panels or instruments.
HVX valves are physically the largest manual valves. A larger port internal diameter allows increased fluid flows even with moderately viscous fluids. It can be used free standing or panel-mounted using an optional locking nut. 6- and 8-port distribution and loop valves are available only in the HVX body style.
Thread depth: 5,6 mm
Threaded connection: 1/4-28