Imprimer
Syringes, Neuros™
Fournisseur: HAMILTON BONADUZ
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
65460-15 65457-01 65458-01 65459-01 65460-02 65460-05 65460-10 65460-20 65457-02 65458-02 65459-02 65460-03 65460-06 65460-11 65460-16 65460-21
549-0703EA 475 EUR
549-0703 549-0697 549-0698 549-0699 549-0700 549-0701 549-0702 549-0704 HAMI65457-02 HAMI65458-02 HAMI65459-02 HAMI65460-03 HAMI65460-06 HAMI65460-11 HAMI65460-16 HAMI65460-21
Syringes, Neuros™
Seringues Seringues de chromatographie
Developed specifically for the neurosciences, these syringes minimise injection site damage by increasing needle rigidity and delivering a micro volume of liquid to an exact location.
  • Compatibility with most infusion pumps and stereotaxic holders
  • Black needle sleeve indicates the exposed needle length
The technology is offered on several models from the 700 Microliter™, 1700 Gastight® and 7000 Modified Microliter™ series. These syringes have very little to no dead volume which preserves the maximum amount of buffer and solution. They feature two different needle sleeves. The sleeve with a blind stop is perfect for cannulated applications and ensures targeted administration with an adjustable penetration depth. The version without a blind stop works best with stereotaxic holders. Both types provide an adjustable needle exposure of 0 to 20 mm.
The 7000 series syringes use a modified Microliter™ plunger which is a tungsten wire that extends the entire length of the needle all the way to the tip, resulting in zero dead volume. This unique design also means if the needle is damaged it cannot be repaired or replaced.
Informations de livraison: Each syringe comes with two needle sleeves. The 7000 series syringes come with the adapter already installed. The adapter kit includes a glass adapter, two needle sleeves and a six-pack of 33 gauge needles.