Imprimer
PIG® Rip-&-Fit® oil-only mat
Fournisseur: NEW PIG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MAT442 MAT443
NEWPMAT442EA 76.3 EUR
NEWPMAT442 NEWPMAT443
PIG® Rip-&-Fit® oil-only mat
Matériaux absorbants
Extra perforations let you create custom sizes of PIG Mat without cutting. Just "rip" it along the easy-tear perfs and "fit" it into place.
  • Extra perforations in our Rip-&-Fit Mat allow you to easily "rip" this PIG Oil-Only Mat and "fit" it to your application without cutting
  • No matter how you rip it, this mat features the proven eight-layer, 100% PP construction of PIG Mat
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave behind liquids or fibre residue
  • Absorbs and retains oils and oil-based liquids - including lubricants and fuels - without taking in a drop of water
  • Bright-white colour makes absorbed oil easier to see; draws attention to machine leaks and clearly shows saturation level during spill response
  • Floats to clean up spills on water
The flame-resistant material won't burn immediately like cellulose mats; melts when exposed to high heat whilst on the job. Perforated every 9.5 cm across width and every 25 cm across length. The heavyweight construction is highly durable and absorbent for demanding tasks and large-volume cleanups.