Imprimer
PIG® Poly utility trays
Fournisseur: NEW PIG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PAK656 PAK657 PAK658 PAK661
NEWPPAK656EA 21.8 EUR
NEWPPAK656 NEWPPAK657 NEWPPAK658 NEWPPAK661
PIG® Poly utility trays
Plateaux/Récipients/Pots Plateaux pour instruments
Flat-bottom poly trays are the economical way to store small parts. Great for lining your benches and shelving..
  • Light-coloured, flat-bottom design makes it easy to see screws, springs and other small parts
  • All-poly construction resists most chemicals; will not rust or corrode
  • Rotationally molded spill tray is ideal for applications that would compromise injected or thermoformed trays
  • Made from FDA-compliant resins for containment of food and cosmetic products
Square modular design allows efficient use of space on bench or shelves for process parts.