Imprimer
Transport trolleys for lab
Fournisseur: Metro International
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CHEMX3 LABX3 MDSX3
139-0059EA 1910 EUR
139-0059 139-0075 139-0076
Transport trolleys for lab
Chariots
Metro’s Lab Cart line includes a General Lab Cart, Chemical Cart, and Mobile Desk. All carts are designed to offer superior corrosion resistance. Smooth rolling, 127 mm polyurethane castors come as standard with each cart. All shelves adjust in 12,7 mm increments, offering precise shelf adjustment and maximum storage utilisation. Microban antimicrobial protects the lower shelf mat from bacteria, mold, mildew and fungi that cause odors, stains and product degradation.
Mobile Desk:
Mobile Desk can be used for instrumentation storage or can be combined with accessories to provide a functional lab work surface. The rear posts are 1524 mm hieght to accommodate the monitor/utility bracket.
Chemical Cart:
Chemical Cart is designed to offer superior corrosion resistance in a lab environment. Stackable perimeter ledges retain chemicals, acid and other solution containers for safe transport. Solid shelf mats lift off of the shelf frame for easy cleaning.
General Lab Cart:
General Lab Cart is designed to offer superior corrosion resistance in a lab environment. Ideal for transporting equipment and instruments in and around the laboratory.