Imprimer
VWR® Separating Funnels, Squibb
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
532-0018EA 68.9 EUR
532-0018 532-0029 532-0019 532-0020 532-0021 532-0022 532-0023 532-0024 532-0025 532-0026 532-0027 532-0028
VWR® Separating Funnels, Squibb
Funnels Separating Funnels
Borosilicate 3.3 glass with NS-PE stopper.
  • With or without graduations
  • Stopcock with PTFE key
  • Very good chemical and temperature resistance