Imprimer
Mixing measuring cylinders, tall form, class A, borosilicate glass, FORTUNA®
Fournisseur: Poulten & Graf
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.4204502 1.4203702
612-0528EA 26.1 EUR
612-0528 612-0530
Mixing measuring cylinders, tall form, class A, borosilicate glass, FORTUNA®
Éprouvettes Éprouvettes graduées
Borosilicate glass 3,3 class A, conformity certified.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, PP stopper
  • Blue graduations
Certifications: DIN EN ISO 4788