Imprimer
Funnels, powder, with short wide stem
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
221-0239EA 30.8 EUR
221-0239 221-0240 221-0241 221-0242
Funnels, powder, with short wide stem
Funnels Powder Funnels
PP, translucent.
  • Parallel stem minimises powder bridging
  • External ribs for better air evacuation
  • Excellent chemical and temperature resistance
  • Autoclavable
Used for transferring powders. Also suitable for liquids.