Imprimer
VWR®, Graduated Pipettes, Type 3, Class B
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
612-4122EA 16.8 EUR
612-4122 612-4123 612-4124 612-4125 612-4126 612-4127 612-4128 612-4121
VWR®, Graduated Pipettes, Type 3, Class B
Pipettes Graduated Pipettes
Borosilicate 9.1 glass.
  • Calibrated to deliver (TD, Ex)
  • Marks and inscriptions in amber stain
  • Cotton plug ending (cotton plugs not included)
DIN 12696
Certifications: DIN 12696