Imprimer
VWR®, Burettes, Straight Stopcock
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
612-4139EA 81.3 EUR
612-4139 612-4140 612-4141
VWR®, Burettes, Straight Stopcock
Burettes
Borosilicate 3.3 glass, class AS.
  • Schellbach stripe, stopcock with glass key
  • High contrast blue enamel graduations
  • Batch certificate included
Certifications: DIN EN ISO 385