Imprimer
PCR buffers
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B34 B33 B38 B16 B55 B65 B71
B34EA 22.65 EUR
B34 B33 B38 B16 B55 B65 B71
PCR buffers
Réactifs pour acides nucléiques End-point PCR Enzymes and Kits
Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA Polymerases (both recombinant and native). Available in different compositions and also without detergents.